Assay Details
| Gene Symbol: ZFP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017969473.1 | XP_017824962.1 | 4 - 5 | 400 | 119 | |
| XM_017969479.1 | XP_017824968.1 | 4 - 5 | 403 | 119 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017969473.1, XM_017969479.1 transcripts.
Set Membership: |
|

ZFP2
Genomic Map