Assay Details
| Gene Symbol: ZFP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017969473.1 | XP_017824962.1 | 5 - 6 | 555 | 70 | |
| XM_017969479.1 | XP_017824968.1 | 5 - 6 | 558 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017969473.1, XM_017969479.1 transcripts.
Set Membership: |
|

ZFP2
Genomic Map