Assay Details
| Gene Symbol: CABYR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003734643.3 | XP_003734691.1 | 2 - 3 | 249 | 133 | |
| XM_008979621.2 | XP_008977869.1 | 2 - 3 | 249 | 133 | ||
| XM_008979623.2 | XP_008977871.1 | 2 - 3 | 214 | 128 | ||
| XM_008979625.2 | XP_008977873.1 | 2 - 3 | 250 | 133 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008979623.2 transcripts.
Set Membership: |
|

CABYR
Genomic Map