Assay Details
| Gene Symbol: TAF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746537.3 | XP_002746583.1 | 1 - 2 | 62 | 68 | |
| XM_008994426.1 | XP_008992674.1 | 1 - 2 | 62 | 68 | ||
More Information
Set Membership: |
|

TAF8
Genomic Map