Assay Details
| Gene Symbol: ESD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002742677.4 | XP_002742723.1 | 6 - 7 | 543 | 85 | |
| XM_008992130.2 | XP_008990378.1 | 6 - 7 | 551 | 85 | ||
| XM_008992134.2 | XP_008990382.1 | 6 - 7 | 578 | 85 | ||
| XM_008992138.1 | XP_008990386.1 | 6 - 7 | 720 | 85 | ||
| XM_008992148.1 | XP_008990396.1 | 6 - 7 | 524 | 85 | ||
More Information
Set Membership: |
|

ESD
Genomic Map