Assay Details
| Gene Symbol: PISD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743698.4 | XP_002743744.1 | 7 - 8 | 1157 | 58 | |
| XM_009009516.2 | XP_009007764.1 | 7 - 8 | 1378 | 58 | ||
| XM_009009521.2 | XP_009007769.1 | 6 - 7 | 905 | 58 | ||
| XM_017979291.1 | XP_017834780.1 | 7 - 8 | 1343 | 58 | ||
| XM_017979292.1 | XP_017834781.1 | 7 - 8 | 1338 | 58 | ||
| XM_017979296.1 | XP_017834785.1 | 8 - 9 | 1528 | 58 | ||
| XM_017979300.1 | XP_017834789.1 | 7 - 8 | 1362 | 58 | ||
| XM_017979302.1 | XP_017834791.1 | 7 - 8 | 1378 | 58 | ||
| XM_017979306.1 | XP_017834795.1 | 9 - 10 | 1435 | 58 | ||
| XM_017979310.1 | XP_017834799.1 | 7 - 8 | 1194 | 58 | ||
More Information
Set Membership: |
|

PISD
Genomic Map