Assay Details
| Gene Symbol: PTGES | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743374.4 | XP_002743420.1 | 2 - 3 | 321 | 99 | |
| XM_017976741.1 | XP_017832230.1 | 2 - 3 | 145 | 99 | ||
More Information
Set Membership: |
|

PTGES
Genomic Map