Assay Details
| Gene Symbol: PBXIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008984494.1 | XP_008982742.1 | 8 - 9 | 1120 | 70 | |
| XM_008984495.1 | XP_008982743.1 | 9 - 10 | 1271 | 70 | ||
| XM_008984496.2 | XP_008982744.1 | 7 - 8 | 869 | 70 | ||
More Information
Set Membership: |
|

PBXIP1
Genomic Map