Assay Details
| Gene Symbol: LOC103792475 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008994603.2 | XP_008992851.1 | 2 - 3 | 3415 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103792475| Gene Symbol: LOC103792475 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008994603.2 | XP_008992851.1 | 2 - 3 | 3415 | 111 | |
Set Membership: |
|