Assay Details
Gene Symbol: CEMIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002742857.3 | XP_002742903.2 | 6 - 7 | 1936 | 64 | |
XM_017973073.1 | XP_017828562.1 | 6 - 7 | 1470 | 64 | ||
XM_017973081.1 | XP_017828570.1 | 6 - 7 | 1657 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
