Assay Details
| Gene Symbol: CFAP100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008982289.2 | XP_008980537.1 | 16 - 17 | 2257 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CFAP100| Gene Symbol: CFAP100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008982289.2 | XP_008980537.1 | 16 - 17 | 2257 | 55 | |
Set Membership: |
|