Assay Details
| Gene Symbol: EFS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009005779.2 | XP_009004027.1 | 6 - 7 | 1933 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EFS| Gene Symbol: EFS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009005779.2 | XP_009004027.1 | 6 - 7 | 1933 | 85 | |
Set Membership: |
|