Assay Details
| Gene Symbol: INKA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758227.4 | XP_002758273.3 | 1 - 2 | 258 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

INKA1| Gene Symbol: INKA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758227.4 | XP_002758273.3 | 1 - 2 | 258 | 85 | |
Set Membership: |
|