Assay Details
| Gene Symbol: AGR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751532.2 | XP_002751578.1 | 5 - 6 | 504 | 66 | |
| XM_002751533.3 | XP_002751579.1 | 5 - 6 | 478 | 66 | ||
More Information
Set Membership: |
|

AGR2
Genomic Map