Assay Details
| Gene Symbol: HMCES | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008982227.2 | XP_008980475.1 | 6 - 7 | 982 | 70 | |
| XM_008982228.2 | XP_008980476.1 | 6 - 7 | 1086 | 70 | ||
| GenBank mRNA | EF215133.1 | - | 1 - 2 | 60 | 70 | |
More Information
Set Membership: |
|

HMCES
Genomic Map