Assay Details
| Gene Symbol: FAM122B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002763285.4 | XP_002763331.2 | 2 - 3 | 347 | 71 | |
| XM_008989925.2 | XP_008988173.1 | 2 - 3 | 347 | 71 | ||
| XM_017968564.1 | XP_017824053.1 | 1 - 2 | 246 | 71 | ||
| XM_017968565.1 | XP_017824054.1 | 1 - 2 | 244 | 71 | ||
| XM_017968566.1 | XP_017824055.1 | 1 - 2 | 245 | 71 | ||
| XM_017968567.1 | XP_017824056.1 | 1 - 2 | 240 | 71 | ||
| XM_017968568.1 | XP_017824057.1 | 1 - 2 | 241 | 71 | ||
More Information
Set Membership: |
|

FAM122B
Genomic Map