Assay Details
| Gene Symbol: CDK5RAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733014.3 | XP_003733062.1 | 2 - 3 | 238 | 63 | |
| XM_008997443.2 | XP_008995691.1 | 2 - 3 | 128 | 63 | ||
| XM_008997444.2 | XP_008995692.1 | 2 - 3 | 199 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008997444.2 transcripts.
Set Membership: |
|

CDK5RAP3
Genomic Map