Assay Details
| Gene Symbol: KYAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009001637.2 | XP_008999885.1 | 13 - 14 | 1739 | 72 | |
| XM_009001639.2 | XP_008999887.1 | 13 - 14 | 1415 | 72 | ||
| XM_009001640.2 | XP_008999888.1 | 14 - 15 | 1428 | 72 | ||
| XM_017974898.1 | XP_017830387.1 | 13 - 14 | 1700 | 72 | ||
More Information
Set Membership: |
|

KYAT3
Genomic Map