Assay Details
| Gene Symbol: C5H17orf80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748696.4 | XP_002748742.1 | 3 - 4 | 1719 | 80 | |
| XM_008997676.2 | XP_008995924.1 | 3 - 4 | 1720 | 80 | ||
| XM_008997677.2 | XP_008995925.1 | 3 - 4 | 1728 | 80 | ||
| XM_008997678.2 | XP_008995926.1 | 3 - 4 | 1734 | 80 | ||
| XM_008997679.2 | XP_008995927.1 | 3 - 4 | 1718 | 80 | ||
| XM_017972782.1 | XP_017828271.1 | 3 - 4 | 1727 | 80 | ||
More Information
Set Membership: |
|

C5H17orf80
Genomic Map