Assay Details
| Gene Symbol: C5H17orf80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748696.4 | XP_002748742.1 | 5 - 6 | 1938 | 74 | |
| XM_017972782.1 | XP_017828271.1 | 5 - 6 | 1946 | 74 | ||
More Information
Set Membership: |
|

C5H17orf80
Genomic Map