Assay Details
| Gene Symbol: PAK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747354.2 | XP_002747400.1 | 7 - 8 | 2212 | 69 | |
| XM_008995745.2 | XP_008993993.1 | 8 - 9 | 2286 | 69 | ||
More Information
Set Membership: |
|

PAK5
Genomic Map