Assay Details
| Gene Symbol: ZNF76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003732648.3 | XP_003732696.1 | 2 - 3 | 284 | 65 | |
| XM_008994297.1 | XP_008992545.1 | 2 - 3 | 292 | 65 | ||
| XM_008994298.2 | XP_008992546.1 | 2 - 3 | 356 | 65 | ||
| XM_008994299.1 | XP_008992547.1 | 2 - 3 | 292 | 65 | ||
| XM_008994300.2 | XP_008992548.1 | 2 - 3 | 372 | 65 | ||
| XM_008994302.1 | XP_008992550.1 | 2 - 3 | 292 | 65 | ||
| XM_008994303.1 | XP_008992551.1 | 2 - 3 | 292 | 65 | ||
| XM_008994304.2 | XP_008992552.1 | 2 - 3 | 292 | 65 | ||
| XM_008994305.2 | XP_008992553.1 | 2 - 3 | 284 | 65 | ||
| XM_008994306.1 | XP_008992554.1 | 2 - 3 | 292 | 65 | ||
| XM_017971031.1 | XP_017826520.1 | 2 - 3 | 509 | 65 | ||
More Information
Set Membership: |
|

ZNF76
Genomic Map