Assay Details
| Gene Symbol: FAM177A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753819.4 | XP_002753865.1 | 5 - 6 | 555 | 93 | |
| XM_009005922.2 | XP_009004170.1 | 5 - 6 | 717 | 93 | ||
| XM_009005923.2 | XP_009004171.1 | 4 - 5 | 642 | 93 | ||
More Information
Set Membership: |
|

FAM177A1
Genomic Map