Assay Details
| Gene Symbol: AP3S2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008998407.2 | XP_008996655.1 | 1 - 2 | 153 | 82 | |
| XM_017973292.1 | XP_017828781.1 | 1 - 2 | 150 | 82 | ||
More Information
Set Membership: |
|

AP3S2
Genomic Map