Assay Details
| Gene Symbol: SPARC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744407.4 | XP_002744453.2 | 7 - 8 | 895 | 69 | |
| XM_008988403.2 | XP_008986651.1 | 7 - 8 | 892 | 69 | ||
More Information
Set Membership: |
|

SPARC
Genomic Map