Assay Details
| Gene Symbol: LOC100398528 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017963576.1 | XP_017819065.1 | 7 - 8 | 738 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100398528| Gene Symbol: LOC100398528 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017963576.1 | XP_017819065.1 | 7 - 8 | 738 | 64 | |
Set Membership: |
|