Assay Details
| Gene Symbol: RDM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008997048.2 | XP_008995296.1 | 4 - 5 | 669 | 71 | |
| XM_008997050.2 | XP_008995298.1 | 3 - 4 | 840 | 71 | ||
| XM_008997052.2 | XP_008995300.1 | 3 - 4 | 489 | 71 | ||
| XM_017972521.1 | XP_017828010.1 | 3 - 4 | 840 | 71 | ||
| XM_017972522.1 | XP_017828011.1 | 3 - 4 | 834 | 71 | ||
| XM_017972523.1 | XP_017828012.1 | 3 - 4 | 840 | 71 | ||
More Information
Set Membership: |
|

RDM1
Genomic Map