Assay Details
| Gene Symbol: HSH2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987463.2 | XP_008985711.2 | 3 - 4 | 598 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HSH2D| Gene Symbol: HSH2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987463.2 | XP_008985711.2 | 3 - 4 | 598 | 70 | |
Set Membership: |
|