Assay Details
| Gene Symbol: CFI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745492.4 | XP_002745538.4 | 1102 | 99 | ||
| XM_017970339.1 | XP_017825828.1 | 1157 | 99 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CFI| Gene Symbol: CFI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745492.4 | XP_002745538.4 | 1102 | 99 | ||
| XM_017970339.1 | XP_017825828.1 | 1157 | 99 | |||
Set Membership: |
|