Assay Details
| Gene Symbol: ZUP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746893.4 | XP_002746939.1 | 9 - 10 | 1949 | 63 | |
| XM_008994946.2 | XP_008993194.1 | 9 - 10 | 1952 | 63 | ||
| XM_017971336.1 | XP_017826825.1 | 9 - 10 | 1871 | 63 | ||
More Information
Set Membership: |
|

ZUP1
Genomic Map