Assay Details
| Gene Symbol: STK35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003732801.3 | XP_003732849.1 | 2 - 3 | 902 | 94 | |
| XM_017971900.1 | XP_017827389.1 | 2 - 3 | 902 | 94 | ||
More Information
Set Membership: |
|

STK35
Genomic Map