Assay Details
| Gene Symbol: RAB11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753256.4 | XP_002753302.1 | 3 - 4 | 718 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB11A| Gene Symbol: RAB11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753256.4 | XP_002753302.1 | 3 - 4 | 718 | 79 | |
Set Membership: |
|