Assay Details
Gene Symbol: SHARPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002759147.4 | XP_002759193.3 | 2 - 3 | 655 | 58 | |
XM_017965517.1 | XP_017821006.1 | 2 - 3 | 655 | 58 | ||
XM_017965518.1 | XP_017821007.1 | 2 - 3 | 655 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
