Assay Details
| Gene Symbol: LIMA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752448.3 | XP_002752494.1 | 7 - 8 | 1145 | 61 | |
| XM_009003760.1 | XP_009002008.1 | 7 - 8 | 1145 | 61 | ||
More Information
Set Membership: |
|

LIMA1
Genomic Map