Assay Details
| Gene Symbol: LOC100395188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760851.4 | XP_002760897.1 | 2 - 3 | 544 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100395188| Gene Symbol: LOC100395188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760851.4 | XP_002760897.1 | 2 - 3 | 544 | 73 | |
Set Membership: |
|