Assay Details
| Gene Symbol: LOC100395188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760851.4 | XP_002760897.1 | 3 - 4 | 613 | 109 | |
| XM_017966688.1 | XP_017822177.1 | 3 - 4 | 288 | 109 | ||
More Information
Set Membership: |
|

LOC100395188
Genomic Map