Assay Details
Gene Symbol: WASHC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002752934.4 | XP_002752980.2 | 12 - 13 | 1111 | 155 | |
XM_009004526.2 | XP_009002774.1 | 11 - 12 | 912 | 155 | ||
XM_017976332.1 | XP_017831821.1 | 7 - 8 | 960 | 155 | ||
XM_017976333.1 | XP_017831822.1 | 5 - 6 | 1108 | 155 |

More Information
Set Membership: |
![]() ![]() |
