Assay Details
| Gene Symbol: WASHC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752934.4 | XP_002752980.2 | 17 - 18 | 1786 | 88 | |
| XM_009004526.2 | XP_009002774.1 | 16 - 17 | 1587 | 88 | ||
| XM_017976332.1 | XP_017831821.1 | 12 - 13 | 1635 | 88 | ||
| XM_017976333.1 | XP_017831822.1 | 10 - 11 | 1783 | 88 | ||
More Information
Set Membership: |
|

WASHC4
Genomic Map