Assay Details
| Gene Symbol: WASHC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752934.4 | XP_002752980.2 | 27 - 28 | 2897 | 120 | |
| XM_009004526.2 | XP_009002774.1 | 26 - 27 | 2698 | 120 | ||
| XM_017976332.1 | XP_017831821.1 | 22 - 23 | 2746 | 120 | ||
| XM_017976333.1 | XP_017831822.1 | 20 - 21 | 2894 | 120 | ||
More Information
Set Membership: |
|

WASHC4
Genomic Map