Assay Details
| Gene Symbol: WASHC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752934.4 | XP_002752980.2 | 2 - 3 | 284 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

WASHC4| Gene Symbol: WASHC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752934.4 | XP_002752980.2 | 2 - 3 | 284 | 108 | |
Set Membership: |
|