Assay Details
| Gene Symbol: SGK494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748246.4 | XP_002748292.2 | 10 - 11 | 1066 | 86 | |
| XM_008997198.2 | XP_008995446.1 | 10 - 11 | 1078 | 86 | ||
| XM_008997199.2 | XP_008995447.1 | 10 - 11 | 1075 | 86 | ||
| XM_008997201.2 | XP_008995449.1 | 9 - 10 | 973 | 86 | ||
More Information
Set Membership: |
|

SGK494
Genomic Map