Assay Details
| Gene Symbol: MRPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745705.4 | XP_002745751.3 | 7 - 8 | 848 | 75 | |
| XM_008993109.2 | XP_008991357.1 | 7 - 8 | 886 | 75 | ||
More Information
Set Membership: |
|

MRPL1
Genomic Map