Assay Details
| Gene Symbol: MRPS24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751363.4 | XP_002751409.1 | 1 - 2 | 111 | 49 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MRPS24| Gene Symbol: MRPS24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751363.4 | XP_002751409.1 | 1 - 2 | 111 | 49 | |
Set Membership: |
|