Assay Details
Gene Symbol: EMC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002761230.4 | XP_002761276.1 | 2 - 3 | 558 | 60 | |
XM_008986327.2 | XP_008984575.1 | 2 - 3 | 622 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
