Assay Details
| Gene Symbol: FUOM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756729.4 | XP_002756775.2 | 4 - 5 | 362 | 90 | |
| XM_008978963.2 | XP_008977211.1 | 4 - 5 | 361 | 90 | ||
More Information
Set Membership: |
|

FUOM
Genomic Map