Assay Details
Gene Symbol: MAP3K7CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002761340.4 | XP_002761386.1 | 4 - 5 | 1138 | 101 | |
XM_008986545.2 | XP_008984793.1 | 4 - 5 | 383 | 101 | ||
XM_017967227.1 | XP_017822716.1 | 2 - 3 | 237 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
