Assay Details
| Gene Symbol: CDH19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757317.3 | XP_002757363.1 | 11 - 12 | 2111 | 93 | |
| XM_008979947.2 | XP_008978195.1 | 11 - 12 | 2064 | 93 | ||
More Information
Set Membership: |
|

CDH19
Genomic Map