Assay Details
| Gene Symbol: GCSAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758818.3 | XP_002758864.1 | 1 - 2 | 255 | 141 | |
| XM_017965252.1 | XP_017820741.1 | 1 - 2 | 255 | 141 | ||
| XM_017965254.1 | XP_017820743.1 | 1 - 2 | 255 | 141 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002758818.3, XM_017965252.1, XM_017965254.1 transcripts.
Set Membership: |
|

GCSAM
Genomic Map