Assay Details
| Gene Symbol: HECA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002806735.3 | XP_002806781.1 | 2 - 3 | 1317 | 90 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

HECA| Gene Symbol: HECA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002806735.3 | XP_002806781.1 | 2 - 3 | 1317 | 90 | |