Assay Details
| Gene Symbol: TEAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988374.2 | XP_008986622.1 | 9 - 10 | 801 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TEAD2| Gene Symbol: TEAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988374.2 | XP_008986622.1 | 9 - 10 | 801 | 56 | |
Set Membership: |
|